Learn More
BPIFB3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA561086
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: VSLGALGSVE FSLATLPLIS NQYIELDINP IVKSVAGDII DFPKSRAPAK VPPKKDHTSQ VMVPLYLFNT TFGL Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.
BPI fold-containing family B member 3 may have the capacity to recognize and bind specific classes of odorants. May act as a carrier molecule, transporting odorants across the mucus layer to access receptor sites. May serve as a primary defense mechanism by recognizing and removing potentially harmful odorants or pathogenic microorganisms from the mucosa or clearing excess odorant from mucus to enable new odorant stimuli to be received.Specifications
BPIFB3 | |
Polyclonal | |
Unconjugated | |
BPIFB3 | |
antimicrobial peptide RYA3; BPI fold containing family B member 3; BPI fold containing family B, member 3; BPI fold-containing family B member 3; Bpifb3; C20orf185; Ligand-binding protein RYA3; long palate, lung and nasal epithelium carcinoma-associated protein 3; Lplunc3; RYA3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
359710 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P59826 | |
BPIFB3 | |
Recombinant protein corresponding to Human BPIFB3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only