missing translation for 'onlineSavingsMsg'
Learn More

Brady Printer IP printer 240mm x 312mm x 435mm label 300dpi

Brand:  Brady 360844

Printer IP printer 240mm x 312mm x 435mm label 300dpi

Code : 03

Additional Details : Weight : 9.00800kg

Product Code. 11573225

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.