missing translation for 'onlineSavingsMsg'
Learn More

BP 554 maleate, Tocris Bioscience™

Selective 5-HT1A agonist

Brand:  Tocris (Bio-Techne) 0556/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13487753

  • £128.25 / 10mg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

Selective 5-HT1A agonist.
Specifications

Chemical Identifiers

1221401-95-1
456.495
1-[3-(3, 4-Methylenedioxyphenoxy)propyl]-4-phenyl-piperazine maleate
C24H28N2O7
NBCXNOHQTALBRA-BTJKTKAUSA-N
56972150

Specifications

BP 554 maleate
1221401-95-1
C24H28N2O7
NBCXNOHQTALBRA-BTJKTKAUSA-N
1-[3-(1,3-benzodioxol-5-yloxy)propyl]-4-phenylpiperazine;(Z)-but-2-enedioic acid
56972150
10mg
>99%
1-[3-(3, 4-Methylenedioxyphenoxy)propyl]-4-phenyl-piperazine maleate
C1CN(CCN1CCCOC2=CC3=C(C=C2)OCO3)C4=CC=CC=C4.C(=CC(=O)O)C(=O)O
456.495
456.48
Videos