missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bovine IL-1 beta, Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RBOIL1BI
Code : PN
Additional Details : Weight : 0.80000kg
Description
Bovine IL-1 beta Recombinant Protein, RBOIL1BI, from Invitrogen.
Interleukin-1 beta (IL-1 beta) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells. IL-1 beta is synthesized in response to inflammatory stimuli as a 31 kDa inactive pro-form that accumulates in the cytosol. Cleavage of pro-IL-1 beta into the active 17 kDa protein requires the activation of inflammasomes, which are multi-protein complexes that respond to pathogens, stress conditions, and other danger signals. Inflammasome activation triggers the processing of the caspase-1 precursor into its active form, which in turn cleaves pro-IL-1 beta. IL-1 beta lacks a signal sequence peptide for classical ER/Golgi pathway and is secreted alongside caspase-1 via an alternate and incompletely understood mechanism. Although IL-1 beta is most often secreted in its active form, secretion of the uncleaved protein may be detectable under some biological conditions. IL-1 beta signals through two receptors, IL-1RI and IL-1RII, both of which are shared with IL-1 alpha. IL-1 beta activity can be moderated by IL-1 Receptor Antagonist (IL-1RA), a protein produced by many cell types that blocks receptor binding through competitive inhibition. IL-1 beta play an important role in innate host defense by triggering the production of other proinflammatory cytokines in target cells and initiating acute-phase responses to infection and injury. Elevated levels of IL-1 beta have been associated with many chronic inflammatory conditions IL-1 beta neutralizing antibodies potential therapeutic value. For dilution, use a buffer containing carrier protein, e.g. PBS with 1% BSA or a neutral media containing 10% FCS and freeze in aliquots at -70°C. Activity: The biological activity of recombinant bovine IL-1-beta was determined by measuring the proliferation of mouse D10.G4.1 cells (Kaye J. et al. (1984) J. Immunol. 133:1339). The ED50 (or 1 unit of activity) for this effect in our laboratories typically falls into the range of 14.1 – 21.1 pg/mL. Expression: Mature bovine IL-1-beta was expressed in Pichia pastoris, KM71H. GenBank™ Accession &Num; P09428, amino acids 114 - 266. For protein verification, the expressed protein was N-terminal sequenced and begins with the following 10 amino acids: *EAEAAPVQSI. *The underlined sequence is due to incomplete cleavage of the alpha-factor secretory signal sequence by the Ste 13 protease. Reconstitute with 0.5 mL of distilled water.Specifications
P09428 | |
PBS with no preservative | |
Bovine IL-1 beta | |
Yeast | |
RUO | |
Bovine IL-1 beta | |
Bovine | |
Bv | |
Recombinant | |
Recombinant |
ELISA, Immunoprecipitation, Western Blot | |
281251 | |
20μg | |
4°C | |
Bovine Interleukin 1 beta IL1B, IL-1beta | |
IL1B | |
Unconjugated | |
Yeast | |
Lyophilized | |
purified |