Learn More
Thermo Scientific™ Bottom-of-the-Bottle™ Solvent Filters
Protect your HPLC system with Thermo Scientific™ Bottom-of-the-Bottle™ Solvent Filters.
£31.33 - £49.00
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10334573
|
Thermo Scientific™
A-302 |
Fit 1/16 in. OD Tube to 1/8 in. OD Plastic Tubing |
£36.35
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10249464
|
Thermo Scientific™
A-302A |
Fit to 1/8 in. OD Plastic Tubing Using 1/8 in. Polypropylene Nut |
£49.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10371065
|
Thermo Scientific™
A-437 |
1/8 in. OD Tubing |
£42.60
Each |
Estimated Shipment: 01-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10651026
|
Thermo Scientific™
A-436 |
3/16 in. OD Plastic Tubing |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Protect your HPLC system from contamination with Thermo Scientific™ Bottom-of-the-Bottle™ Solvent Filters. The flat design ensures efficient draw. They are made from 100% PEEK polymer and include two 10µm filters. A helium sparge port and frit are incorporated for optional use.
- Efficient draw
- 100% PTFE polymer, including 2μm filter
- Built-in helium sparge port and frit
- Stainless-steel 10μm inlet filters for longer lifetime
Specifications
Filter Kit |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.