missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Bottletop Vacuum Filtration Units

Designed for sterilization or clarification of aqueous cell culture media. Duran™ Bottletop Vacuum Filtration Units include raised and moulded graduation marks for easy volume assessment..

Brand:  Duran™ 292702842

Additional Details : Weight : 2.20000kg

 View more versions of this product

Product Code. 15503557

  • £108.60 / Pack of 12


Estimated Shipment
15-05-2018
Add to basket

Description

Description

For use with 45mm media bottles.
  • Available in three different asymmetric pore sizes: 0.1μm, 0.2μm, or 0.45 μm.
  • Raised moulded graduation marks for easy volume reading.
  • Manufactured in a Class 100,000 clean room from Class VI, non-cytotoxic materials.
  • Supplied sterile.
Specifications

Specifications

Yes
0.45μm
90mm
Yes
PES
90mm
GL45
12Pack
Filter funnel
Duran™ TILT GL 45 Media Bottle
500mL
Videos
Documentation

Technical Documents