missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ TILT Bottle Top Filter Unit, GL 45

Bottle Top Filter Unit with GL 45 thread.

Brand:  DWK Life Sciences 292702826

Additional Details : Weight : 2.20000kg

 View more versions of this product

Product Code. 15593547

  • £147.54 / Pack of 12


Estimated Shipment
27-10-2020
Add to basket

Description

Description

Designed for sterilization or clarification of aqueous cell culture media Supplied as a filter funnel unit for use with the DURAN™ TILT bottle (with GL 45 thread adaptor) or 45 mm media bottles. Comes in three different asymmetric pore sizes (0.1 µm, 0.2 µm, or 0.45 µm). Raised moulded graduation marks for easy volume reading. Manufactured in a Class 100,000 clean room from Class VI, non-cytotoxic materials. Supplied sterile.

  • Low bottle height for easy handling in safety cabinets
  • The raised volume marking allows a clear and distinct reading of the contained quantity at any time
  • Comes in three different asymmetric pore sizes (0.1 µm, 0.2 µm, or 0.45 µm)
  • Manufactured in a Class 100,000 clean room from Class VI, non-cytotoxic materials
  • Supplied sterile
Specifications

Specifications

PES 0.2 µm
500 mL
Boro 3.3
Storage of Samples and Reagents; Sample Collection; Transport and Shipping
92 mm
12/Pk.
GL 45
Videos
Documents