Learn More
Sartorius Bottled Media for Sartorius Membrane Filtration Units
Lysine agar media for wild yeasts determination
Brand: Sartorius 14143----------A
Code : NEW
Additional Details : Weight : 0.01000kg
Description
The traditional culture media for microorganisms is agar media. This can be used for the membrane filtration method or for direct incubation. Agar media in bottles are the cost-effective alternative for casting plates.
- 250mL bottles, 4 bottles per box
- Economical, provides time and cost savings
- According to international formulas
- Consistent quality
Specifications
Liquid | |
Lysine | |
Membrane filtration method or for direct incubation, for determination of wild Yeasts | |
250 mL |
Germany | |
250 mL | |
Bottle |