missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Autoclavable Bottletop Dispensers
Brand: Fisherbrand™
Code : 41
Additional Details : Weight : 0.61673kg
Description
- Wetted parts are PTFE, FEP and borosilicate glass (allows instrument to be used with a wide variety of chemicals)
- 7 volume ranges to choose from
- Bubble-free dispensing
- Effortless plunger movement
- Disassembles for easy cleaning and servicing
- Autoclavable at 121°C, 15 psi for 10 to 15 minutes
- Each bottle top dispenser includes one 28mm, 32mm, 36mm, 40mm, 45mm bottle neck adaptor
Component Description
- Piston: PTFE
- Cylinder: Glass
- Volume Adjustment Knob: Screw Type
- Valve Housing: PTFE
- Valve Assembly: Glass Ball & Seal
- Discharge Assembly: PTFE
- Delivery Tube: FEP
- Inlet Tube: FEP
- Calibration: Individually calibrated and certified (in-lab by user is also possible)
- Accuracy & Reproducibility: In accordance with ISO 8655 standards
- Compability: Excellent with most reagents