missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Bottle Shape With Assembly Set Filtering Flask

Ideal for vacuum filtration.

Brand:  Duran™ 211946854

Code : A0

Additional Details : Weight : 2.19000kg

 View more versions of this product

Product Code. 10169420

  • £134.00 / Each


Estimated Shipment
10-05-2018
Add to basket

Description

Description

With polypropylene plastic hose connection and plastic tubulature. Ideal for vacuum filtration.
Specifications

Specifications

Yes
170mm
Borosilicate Glass
USP Standard
3L
295mm
Videos
Documentation View accessory products