missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Bottle Shape With Assembly Set Filtering Flask

Ideal for vacuum filtration.

Brand:  Duran™ 211947353

Code : A0

Additional Details : Weight : 2.94000kg

 View more versions of this product

Product Code. 10768511

  • £189.00 / Each


Estimated Shipment
11-06-2018
Add to basket

Description

Description

With polypropylene plastic hose connection and plastic tubulature. Ideal for vacuum filtration.
Specifications

Specifications

Yes
185mm
Borosilicate Glass
USP Standard
5000mL
360mm
Videos
Documentation View accessory products