missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Bottle Shape Glass Hose Connection Filtering Flask

Brand:  Duran™ 211916802

Code : A0

Additional Details : Weight : 2.05700kg

 View more versions of this product

Product Code. 10736981

  • £133.00 / Each


Estimated Shipment
08-03-2018
Add to basket

Specifications

Specifications

Yes
170mm
Borosilicate Glass
USP Standard
3L
295mm
Videos