missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Bottle Shape Glass Hose Connection Filtering Flask

Brand:  Duran™ 211918803

Code : A0

Additional Details : Weight : 6.43700kg

 View more versions of this product

Product Code. 10263261

  • £383.00 / Each


Estimated Shipment
31-01-2018
Add to basket

Specifications

Specifications

Yes
255mm
Borosilicate Glass
USP Standard
15000mL
500mm
Videos