missing translation for 'onlineSavingsMsg'
Learn More

Poulten Graf™ Borosilicate Serological Pipet

Brand:  Poulten Graf™ D802

Code : 02

Additional Details : Weight : 0.80000kg

Product Code. 11758245

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Specifications

Specifications

Borosilicate Glass
0.1 mL
1 mL
Serological Pipette
Yes
500/Pk.
270 mm
Single-use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers