Learn More
Karl Hecht™ Volumetric Flask with PP Stopper
Simply and accurately measure required sample volumes. Karl Hecht™ Volumetric Flask with PP Stopper is thick-walled, clearly marked and labeled, and has a solid finish.
£25.02 - £28.99
Product Code | Brand | Capacity (Metric) | Closure Size | Neck Style | Tolerance | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Size | Neck Style | Tolerance | Price | Quantity & Availability | ||
10608671
|
Karl Hecht™
42094068 |
300 mL | NS 14 | Standard | 0.23 mL |
£25.02
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
||
10618671
|
Karl Hecht™
42094072 |
400 mL | NS 19 | Standard | 0.38 mL |
£28.99
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
||
Description
- Duran™ borosilicate glass 3.3.
- Standard type.
- Fused blue graduation.
- Standard ground and interchangeable NS stopper of polyethylene.
- Thick-walled, solid finish.
- Clearly legible figures and marks.
Specifications
Yes |