Learn More
Duran™ Borosilicate Glass Type 3D Filter Funnels
Made of borosilicate glass. DURAN™ Borosilicate Glass Type 3D Filter Funnels are used for qualitative inorganic analysis and preparative chemistry.
£36.15 - £65.90
Specifications
Capacity (Metric) 50 mL
Diameter (Metric) Disc 35mm
Material Borosilicate Glass
Product Type Filter funnel
View More Specs
Products 5
Product Code Brand Porosity Price Quantity & Availability  
Product Code Brand Porosity Price Quantity & Availability  
10747171 View Documents Duran™
258520307
3 £36.15
Each View alternative products
Save Up to 
»
Estimated Shipment
26-03-2020
Add to basket
10424461 View Documents Duran™
258520401
4 £41.80
Each View alternative products
Save Up to 
»
Estimated Shipment
26-03-2020
Add to basket
10028000 View Documents Duran™
258520204
2 £40.35
Each View alternative products
Save Up to 
»
Estimated Shipment
26-03-2020
Add to basket
10304021 View Documents Duran™
258520504
5 £65.90
Each View alternative products
Save Up to 
»
Estimated Shipment
26-03-2020
Add to basket
10706981 View Documents Duran™
258520101
1 £36.15
Each View alternative products
Save Up to 
»
Estimated Shipment
26-03-2020
Add to basket
Description
Description
  • Good thermal shock and chemical resistance 
  • Mate to the filtering flask via a conical rubber seal (GUKO)
Specifications
Specifications
50 mL
35mm
Borosilicate Glass
Filter funnel