missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Borosilicate Glass Type 3D Filter Funnels

Made of borosilicate glass. DURAN™ Borosilicate Glass Type 3D Filter Funnels are used for qualitative inorganic analysis and preparative chemistry.

Brand:  Duran™ 258520504

Code : A0

Additional Details : Weight : 0.04000kg

 View more versions of this product

Product Code. 10304021

  • £59.80 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Good thermal shock and chemical resistance 
  • Mate to the filtering flask via a conical rubber seal (GUKO)
Specifications

Specifications

35mm
Filter funnel
Borosilicate Glass
5
50mL
Videos