Learn More
Borosilicate Glass Stopcock Adapter with Cone
Control flow rate. Quickfit Jointed Ware™ QuickFit™ borosilicate glass stopcock adater with cone comes with a GP Rotoflo™ stopcock.
Brand:  Quickfit MF11/2/SC
74.12 GBP valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Code : 01
Additional Details : Weight : 0.04140kg
 View more versions of this product
Product Code. 11302045
  • £123.00 / Each
Estimated Shipment: 16-05-2024
to see stock.
Add to basket
Description
Description
Control flow rate. The QuickFit™ borosilicate glass stopcock adater with cone comes with a GP Rotoflo™ stopcock.
  • With GP Rotaflo™ PTFE stopcock: - No lubrication required - No contaminating greases - Fine flow control - Positive closure
  • Detachable plastic screw thread connectors allow rubber hose etc. , to be fitted easily and safely
Specifications
Specifications
Stopcock Adapter
Borosilicate Glass
19/26
Cone, screw thread adapter and GP Rotaflo™ PTFE stopcock