Learn More
GPE Scientific™ Borosilicate Glass Stirrers
For use with overhead stirrer motors
Brand:  GPE Scientific™ 30504
Code : 03
Additional Details : Weight : 0.12500kg
Product Code. 12997066
  • £110.00 / Each
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
Description
Description
  • For use with overhead stirrer motors
  • Non-whip and autoclavable
  • The PTFE blades are easily removed or replaced
Specifications
Specifications
1 Pack
Stirrer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products