missing translation for 'onlineSavingsMsg'
Learn More

Robu™ Borosilicate Glass Standard Filter Crucibles

Brand:  Robu™ 20305

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10032151

  • £19.80 / Each


Estimated Shipment
19-03-2020
Add to basket

Specifications

Specifications

Borosilicate Glass
36mm
Analytical filtration in laboratories
5
30mL
36mm
56mm
Standard Filter Crucible
Videos
Documents