Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Squibb Separating Funnels with Solid Glass Plug, Scale
For use in liquid-liquid extractions to separate components. Lenz Laborglasinstrumente™ Borosilicate Glass Squibb Separating Funnels with Solid Glass Plug, Scale are manufactured from durable DURAN™ glass.
Brand: Lenz Laborglasinstrumente™ 04012049
Code : A0
Additional Details : Weight : 0.32400kg
Description
- With NS socket and PE stopper
- Scale of approx. volume/mL
- With screwthread retaining device
Specifications
250 mL | |
Borosilicate Glass | |
2Pack | |
Polyethylene |