missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poulten Graf™ Borosilicate Glass Serological Pipette
Offering economical single-use convenience leading to greatly reduced possibility of cross-contamination. Poulten Graf™ Borosilicate Glass Serological Pipettes are disposable and have clear advantages over plastic counterparts in areas of chemical and temperature resistance, accuracy, transparency and clarity of markings.
£231.00 - £293.00
Specifications
Material | Borosilicate Glass |
---|---|
Graduated | Yes |
Type | Serological Pipette |
Disposable | Single-use |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
11798245
|
Poulten Graf™
D803/PS |
2 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11708255
|
Poulten Graf™
D808 |
10 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Ideal for use with radioactive and other hazardous materials
- Volumes are color-coded
- Environmentally friendly compared with plastic counterparts
Specifications
Borosilicate Glass | |
Yes | |
Serological Pipette | |
Single-use |