Learn More
Day Impex™ Borosilicate Glass Refill
Designed with practicality and safety in mind. Day Impex™ Borosilicate Glass Refill is ultra-low evacuated to allow for safe and efficient handling of all liquified gases, freezing mixtures, and specimens.
Brand: Day Impex™ 210011
Code : 03
Additional Details : Weight : 1.50000kg
Description
- Borosilicate glass inner vessels
Specifications
150 mm | |
For use with Stainless Steel and Enamelled Steel container | |
280 mm | |
4500 mL |
6 in. | |
280mm | |
Borosilicate Glass |