missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ Borosilicate Glass/PTFE Gastight Syringes

Brand:  Hamilton™ 201400

Code : A0

Additional Details : Weight : 0.16500kg

 View more versions of this product

Product Code. 10512131

  • £213.36 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

No
1000 series diluter/dispensers
PTFE
50°C
20°C
AccuDil (AD)
25mL
Liquid or gas sampling
200psi
1025
1/Ea.
Gastight Syringe
Videos