Learn More
Lenz Laborglasinstrumente™ Flat Bottom Hollow Stoppers, Hexagonal Plate
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus. Lenz Laborglasinstrumente™ Flat Bottom Hollow Stoppers, Hexagonal Plate are manufactured from durable DURAN™ borosilicate glass.
Brand: Lenz Laborglasinstrumente™ 01400324
Code : A0
Additional Details : Weight : 0.80000kg
Description
- Available in a range of cone sizes
- Corrosion-free and resistant to thermal shock
Specifications
Hollow stopper | |
10/Pk. | |
Borosilicate Glass | |
NS 24/29 |