missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Dropping Bottle, Amber, Soda-Lime Glass, Supplied with ground jointed pipette, Complete with Rubber teat
134.73 GBP valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Use promo code "23258" to get your promotional price.
Click to view available options
Capacity (Metric):
50 mL
100 mL
Description
Dropping bottles are suitable for liquids which need to be dispensed in small amounts, especially drop by drop. Amber glass bottle provides protection from light for photosensitive reagents. Supplied with rubber teat and clear ground jointed dropping pipette. Replacement clear glass pipettes and natural rubber teats are available.
- Amber glass bottle provides protection from light for photosensitive reagents
- Clear glass pipette with standard ground joint
- Tight closure due to standard taper ground joint
- Natural rubber teat
Specifications
Capacity (Metric) | 100 mL |
Description | From borosilicate 3.3 |
For Use With (Application) | Storage and dispensing of photosensitive liquids and reagents |
Material | Soda Lime Glass |
Quantity | 10/Pk. |
Autoclavable | Not Autoclavable |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'