Learn More
BRAND™ BLAUBRAND™ Borosilicate Glass Calibrated Density Bottles
Calibrated density bottle made with Borosilicate glass that is ideal for use in most research, scientific, and other types of laboratory environments.
Brand: BRAND™ 43320
53.85 GBP valid until 2024-03-30
Use promo code "21741" to get your promotional price.
Additional Details : Weight : 0.03960kg
Description
Gay-Lussac density bottles manufactured from borosilicate glass.- Conforms to DIN ISO 3507 standards
- Calibrated to contain (TC, In).
- Includes individual certificate.
- Stopper NS 10/19 with capillary.
- Top of stopper ground and polished.
- The volume in cm³ is specified to a precision of 3 decimal places.
Specifications
1Each | |
25 mL | |
Pycnometer |
Gay-Lussac | |
Borosilicate Glass |