missing translation for 'onlineSavingsMsg'
Learn More

Conical Filter Vacuum Flask with Hose Tubulature

Use this Conical Filter Vacuum Flask with Hose Tubulature with a vacuum funnel and filter. This thick-walled Ehlenmeyer flask features a glass tube protrusion about an inch from its neck and is also known as a Büchner flask.

Brand:  N/A FHD-345-070S

Code : 01

Additional Details : Weight : 1.50000kg

 View more versions of this product

Product Code. 12353886

Quantity Price
1 £34.80 / Each
10 £31.32 / Each
In Stock 11

Add to basket


Description

Description

  • With 8mm outer diameter tubulature
  • Thick-walled borosilicate glass
  • Vacuum flask
Specifications

Specifications

8mm
500mL
Borosilicate Glass
Videos
Documentation