missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conical Filter Vacuum Flask with Hose Tubulature
Use this Conical Filter Vacuum Flask with Hose Tubulature with a vacuum funnel and filter. This thick-walled Ehlenmeyer flask features a glass tube protrusion about an inch from its neck and is also known as a Büchner flask.
£57.80 - £148.00
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
12363886
|
N/A
2418/1000 |
1000 mL |
£119.00
Each |
In Stock
Log in to see stock. |
|||||
12373886
|
N/A
FHD-345-110J |
2000 mL |
£148.00
Each |
In Stock
Log in to see stock. |
|||||
12343886
|
N/A
FHD-345-050B |
250 mL |
£57.80
Each |
In Stock
Log in to see stock. |
|||||
12353886
|
N/A
FHD-345-070S |
500 mL |
£102.00
Each |
In Stock
Log in to see stock. |
|||||
Description
- With 8mm outer diameter tubulature
- Thick-walled borosilicate glass
- Vacuum flask
Specifications
8 mm | |
Borosilicate Glass |