missing translation for 'onlineSavingsMsg'
Learn More

Robu™ Borosilicate Glass Buchner Filter Funnels

Brand:  Robu™ 21753

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10048121

  • £22.65 / Each


Estimated Shipment
21-02-2020
Add to basket

Specifications

Specifications

Borosilicate Glass
10mm
Highly resistant to most chemicals such as solvents, acids and alkalis
Buchner filter funnel
50 mm
52mm
52mm (OD); 48mm (ID)
3
75 mL
Videos
Documents