missing translation for 'onlineSavingsMsg'
Learn More

Robu™ Borosilicate Glass Buchner Filter Funnels

Brand:  Robu™ 21253

Additional Details : Weight : 0.27000kg

 View more versions of this product

Product Code. 10151020

  • £48.83 / Each


In stock, ready to ship within 16 to 18 days: 8
Add to basket

Description

Description

Specifications

Specifications

85mm (OD); 80mm (ID)
18mm
65 mm
85mm
Buchner filter funnel
250 mL
Borosilicate Glass
Highly resistant to most chemicals such as solvents, acids and alkalis
3
Videos
SDS
Documents
Certificates
View accessory products