missing translation for 'onlineSavingsMsg'
Learn More

Robu™ Borosilicate Glass Buchner Filter Funnels

Brand:  Robu™ 21253

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10003154

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Specifications

Specifications

85mm
85mm (OD); 80mm (ID)
3
250 mL
Borosilicate Glass
18mm
Highly resistant to most chemicals such as solvents, acids and alkalis
Buchner filter funnel
65 mm
Videos
SDS
Documents