missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Borosilicate Glass Blanks

Brand:  Duran™ 210373408

Code : A0

Additional Details : Weight : 1.17000kg

 View more versions of this product

Product Code. 10621682

  • £170.00 / Pack of 6


Estimated Shipment
29-01-2018
Add to basket

Specifications

Specifications

100mm
Borosilicate Glass
42mm
Blank
Videos