missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Borosilicate Glass 3.3 Vaccum Desiccators

Drying of moist samples and storage of moisture-sensitive substances

Brand:  Duran™ 247814603

Code : A0

Additional Details : Weight : 1.11000kg

 View more versions of this product

Product Code. 10539821

  • £135.00 / Each


Estimated Shipment
22-02-2018
Add to basket

Description

Description

  • Made from DURAN™ borosilicate glass
  • Components are vacuum tight (DIN 12491) 
  • Spare parts such as lids and bases can be interchanged (observe DN)
  • NOVUS standard ground joint (NS 24/29)
Specifications

Specifications

∽0.7L
187mm
Borosilicate Glass 3.3
153 ±2mm flange
DIN 12491
Flat Flange And Knobbed Lid
119mm flange
Vaccum Desiccator
Videos
View accessory products