missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Borosilicate Glass 3.3 Complete Vacuum Desiccator

Features NOVUS standard ground joint (NS 24/29) junction tube in lid, Duran™ Borosilicate Glass 3.3 Complete Vacuum Desiccator also includes stopcock and flat flange.

Brand:  Duran™ 247826157

Code : NEW

Additional Details : Weight : 5.48050kg

 View more versions of this product

Product Code. 13435499

  • £329.00 / Each


In Stock 2
Add to basket

Description

Description

  • Vacuum tight
  • Borosilicate glass 3.3
  • Vacuum applied via stopcock
  • Interchangeable spare parts i.e., lids, bases, stopcocks and caps
Specifications

Specifications

∽5.8L
Tap, And Lid
222mm flange
296mm
Vaccum Desiccator
DIN 12491
DN 200 (nominal diameter)
270 ± 2mm flange
Borosilicate Glass 3.3
Videos
Documents