Learn More
Duran™ Borosilicate Desiccator Base with Flat Flange
Designed for use under full vacuum. DURAN™ Borosilicate Desiccator Base with Flat Flange is DIN 12419 certified.
£82.50 - £406.00
Specifications
Material Duran Borosilicate
For Use With (Application) all lid types
Product Type Desiccator Base
View More Specs
Products 5
Product Code Brand Diameter (Metric) Outer Height (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Outer Height (Metric) Price Quantity & Availability  
10576752 View Documents Duran™
247704604
153mm flange 112mm £82.50
Each View alternative products
Save Up to 
»
Estimated Shipment
06-05-2020
Add to basket
10299080 View Documents Duran™
247706605
320mm flange 235mm £239.00
Each View alternative products
Save Up to 
»
Estimated Shipment
06-05-2020
Add to basket
10300411 View Documents Duran™
247706905
380mm flange 283mm £406.00
Each View alternative products
Save Up to 
»
Estimated Shipment
29-04-2020
Add to basket
10772552 View Documents Duran™
247705703
215mm flange 154mm £85.20
Each View alternative products
Save Up to 
»
Estimated Shipment
06-05-2020
Add to basket
10025250 View Documents Duran™
247706108
270mm flange 202mm £97.00
Each View alternative products
Save Up to 
»
Estimated Shipment
06-05-2020
Add to basket
Description
Description
  • DIN 12491
  • Typical applications: drying of moist products and storage of moisture-sensitive substances.
  • Used for drying moist substances or as storage vessels for moisture-sensitive products
  • Designed for use under full vacuum
Specifications
Specifications
Duran Borosilicate
all lid types
Desiccator Base