missing translation for 'onlineSavingsMsg'
Learn More

Duran™ BoroSilicate Crystallizing Dishes with Spout

Includes spout. DURAN™ BoroSilicate Crystallizing Dishes with Spout have good thermal shock resistance.

Brand:  Duran™ 213116309

Code : A0

Additional Details : Weight : 0.65800kg

 View more versions of this product

Product Code. 10599831

  • £89.25 / Each


In Stock 18
Add to basket

Specifications

Specifications

3500mL
230mm
Duran BoroSilicate
230mm
100mm
Crystallizing Dish
Videos