missing translation for 'onlineSavingsMsg'
Learn More

Duran™ BoroSilicate Crystallizing Dishes

Without spout or print. DURAN™ BoroSilicate Crystallizing Dishes have good thermal shock resistance.

Brand:  Duran™ 213133802

Code : A0

Additional Details : Weight : 0.45000kg

 View more versions of this product

Product Code. 11776582

  • £91.70 / Pack of 10


Estimated Shipment
14-06-2018
Add to basket

Specifications

Specifications

100mL
70mm
Duran BoroSilicate
70mm
40mm
Crystallizing Dish
Videos