missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boron trifluoride dimethyl etherate, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 mL
250 mL
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
Specifications
Melting Point | -14.0°C |
Color | Brown |
Density | 1.2300g/mL |
Boiling Point | 126.0°C to 127.0°C |
Flash Point | 62°C |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Linear Formula | (CH3)2O·BF3 |
Refractive Index | 1.3070 to 1.3090 |
Quantity | 50 mL |
Show More |
Safety and Handling
Product Identifier
- Boron trifluoride dimethyl etherate
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 1
- Substance which\, in contact with water\, emits flammable gases Category 1
- H318-Causes serious eye damage.
- H372-Causes damage to organs.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- EUH014-Reacts violently with water.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'