missing translation for 'onlineSavingsMsg'
Learn More

Boron tribromide, 99% min, Thermo Scientific Chemicals

Catalog Number p-7050203
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
10294-33-4
BBr3
250.52
MFCD00011312
ILAHWRKJUDSMFH-UHFFFAOYSA-N
boron tribromide, borane, tribromo, boron bromide, tribromoboron, hsdb 327, tribromoboran, boron tribrornide, boron tri bromide, boron tri-bromide
25134
tribromoborane
BrB(Br)Br
This item is not returnable. View return policy
10294-33-4
BBr3
250.52
MFCD00011312
ILAHWRKJUDSMFH-UHFFFAOYSA-N
boron tribromide, borane, tribromo, boron bromide, tribromoboron, hsdb 327, tribromoboran, boron tribrornide, boron tri bromide, boron tri-bromide
25134
tribromoborane
BrB(Br)Br

As a catalyst in the manufacturing of diborane, ultra pure boron

As a catalyst in the manufacturing of diborane, ultra pure boronBoron tribromide is used as a reactant for luminescent polystyrene derivatives with sterically protected carbazolylborane moieties, high-quality boron-doped graphene via

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 90°C
Melting Point -46°C
Color Colorless to Orange
Physical Form Liquid
Quantity 50 g
UN Number UN2692
Merck Index 14,1347
Solubility Information Miscible with ethanol and carbon tetrachloride.
Formula Weight 250.52
Percent Purity ≤99%
Sensitivity Moisture sensitive
Density 2.643 g/mL
Chemical Name or Material Boron tribromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'