missing translation for 'onlineSavingsMsg'
Learn More

Boron powder, crystalline, -4+40 mesh, Puratronic™, 99.9999% (metals basis), Thermo Scientific Chemicals

Catalog Number 11301168
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7440-42-8
B
10.81
MFCD00134034 MFCD00151272
ZOXJGFHDIHLPTG-UHFFFAOYSA-N
boron, metallic, unii-n9e3x5056q, boron, amorphous, powder, crystalline, boro, boron-elemental, boron, elemental, and compounds, boracium, boranylidyne
5462311
CHEBI:27560
boron
[B]
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7440-42-8
B
10.81
MFCD00134034 MFCD00151272
ZOXJGFHDIHLPTG-UHFFFAOYSA-N
boron, metallic, unii-n9e3x5056q, boron, amorphous, powder, crystalline, boro, boron-elemental, boron, elemental, and compounds, boracium, boranylidyne
5462311
CHEBI:27560
boron
[B]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
UN Number UN3178
Quantity 1 g
Percent Purity 99.9999%
Grade Puratronic™
Mesh Size −4+40 Mesh
Odor Odorless
Form Crystalline Powder
Assay (metals basis)
Chemical Name or Material Boron powder
compliance-icons compliance-icons
Product Identifier
  • Boron powder, crystalline, Puratronic
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3
Hazard Statement
  • H228-Flammable solid.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P264-Wash thoroughly after handling.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'