missing translation for 'onlineSavingsMsg'
Learn More

Boron oxide, Puratronic™, (H{2}O 200ppm), 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number 11306188
Click to view available options
Quantity:
1 cyl
10 x 55 g
This item is not returnable. View return policy
1303-86-2
B2O3
69.62
MFCD00011315
JKWMSGQKBLHBQQ-UHFFFAOYSA-N
boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g
518682
CHEBI:30163
oxo(oxoboranyloxy)borane
O=BOB=O
This item is not returnable. View return policy
1303-86-2
B2O3
69.62
MFCD00011315
JKWMSGQKBLHBQQ-UHFFFAOYSA-N
boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g
518682
CHEBI:30163
oxo(oxoboranyloxy)borane
O=BOB=O

In the electronics industry as a liquid encapsulent for Czochralski-type growth of III-V semiconductor single crystals (e.g., GaAs, GaP, InP).Boron oxide is used as a flux, an acid catalyst and a chemical intermediate in the production of boron halides, esters, carbide, nitride and metallic borides. It has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from talc. It is actively involved in the production of borosilicate glass. It is an additive used in high-temperature lubricants, heatproof glasswares and dopants in semiconductor materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In the electronics industry as a liquid encapsulent for Czochralski-type growth of III-V semiconductor single crystals (e.g., GaAs, GaP, InP)
TRUSTED_SUSTAINABILITY
Boiling Point 1860°C
Melting Point ∼450°C
Color White
Physical Form Cylinder
Quantity 1 cyl
Merck Index 14,1337
Solubility Information Slightly soluble in water and methanol.
Formula Weight 69.62
Percent Purity 99.999%
Grade Puratronic™
Odor Odorless
Sensitivity Hygroscopic
Density 2.46 g/mL (±)0.01
Assay (metals basis)
Chemical Name or Material Boron oxide, H2O 200ppm, Cylinder, 5.0cm dia.
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'