missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boron oxide, 98%, -40 mesh, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Glass Bottle
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 1860.0°C |
Melting Point | 450.0°C |
Color | White |
Physical Form | Powder |
Quantity | 25 kg |
Assay Percent Range | 98% |
Linear Formula | B2O3 |
Fieser | 01,673; 02,32; 03,33; 09,1349 |
Merck Index | 15,1348 |
Solubility Information | Solubility in water: 36g/L (25°C). Other solubilities: soluble in hot water, soluble in alcohol, glycerol and acids |
Show More |
Safety and Handling
Product Identifier
- Boron oxide
- Danger
- Reproductive toxicity Category 1B
- H360FD-May damage fertility. May damage the unborn child.
- P201-Obtain special instructions before use.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'