missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Boron nitride sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.99% (metals basis)

Brand:  Alfa Aesar™ 041053.KS

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15423775

  • £798.39 / Each


Estimated Shipment date
from Supplier 11-05-2018
Add to basket

Specifications

Specifications

Boron nitride sputtering target
99.99%
24.82
Odorless
50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
(metals basis)
MFCD00011317
1 each
Videos
SDS