missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Boron nitride sputtering target, 25.4mm (1.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.99% (metals basis excluding Ca)

Brand:  Alfa Aesar™ 044876.KS

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 1 each

Product Code. 15433915

  • £306.34 / Each


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Specifications

Boron nitride sputtering target
99.99%
BN
MFCD00011317
1 each
25.4mm (1.0 in.) dia. x 3.18mm (0.125 in.) thick
(metals basis excluding Ca)
24.82
Odorless
Videos
SDS