missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Boron nitride, Refractory Brushable Paint, BN 23%

Brand:  Alfa Aesar™ 043279.A1

Code : NEW

Additional Details : CAS Number : 10043-11-5 Weight : 1.00000kg

Quantity 1kg

 View more versions of this product

Product Code. 11359398

  • £88.40 / 1kg


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

Boron nitride
Refractory Brushable Paint
MFCD00011317
10043-11-5
BN

Specifications

Boron nitride
10043-11-5
BN
BN 23%
White
Refractory Brushable Paint
Paste
MFCD00011317
Odorless
1kg
Videos
SDS