missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Boron nitride, low binder, 99.5% (metals basis)

Brand:  Alfa Aesar™ 043254.KJ

Code : NEW

Additional Details : CAS Number : 10043-11-5 Weight : 0.01000kg

Quantity 5 piece

 View more versions of this product

Product Code. 15491877

  • £99999.00 / Pack of 5


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

Boron nitride
Low binder, 9.5mm dia. x 406mm long
24.82
14,1346
10043-11-5
BN
MFCD00011317

Specifications

Boron nitride
10043-11-5
99.5%
BN
MFCD00011317
Odorless
Low binder, 9.5mm dia. x 406mm long
Hot Pressed Rod
(metals basis)
24.82
14,1346
5 piece
Videos
SDS