missing translation for 'onlineSavingsMsg'
Learn More

Boron Nitride Bar, L x W x H: 300 x 6.4 x 6.4mm, Thermo Scientific™

Catalog Number 15456995
Click to view available options
Quantity:
1 Pc.
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 10043-11-5
Physical Form Bar
Quantity 1 Pc.
Molecular Formula BN
MDL Number MFCD00011317
Synonym boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r
InChI Key LNLSXDSWJBUPHM-UHFFFAOYSA-N
SMILES B=N
IUPAC Name boranimine
Molecular Weight (g/mol) 24.82
PubChem CID 66227
ChEBI CHEBI:50883
Formula Weight 24.82
Odor Odorless
Chemical Name or Material Boron nitride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'