missing translation for 'onlineSavingsMsg'
Learn More

Boron nitride, 99.5% (metals basis), Alfa Aesar™

Used in production of alloys. It is also used in the manufacture of furnace refractories, insulation materials, machinery and aviation.

Brand:  Alfa Aesar 044837.KJ

Code : NEW

Additional Details : CAS Number : 10043-11-5 Weight : 0.01000kg

Quantity 5 pc.

 View more versions of this product

Product Code. 15402047

  • £927.00 / Pack of 5

Estimated Shipment: 26-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

10043-11-5
24.82
PZNSFCLAULLKQX-UHFFFAOYSA-N
66227
boranylidyneamine
BN
MFCD00011317
boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r
CHEBI:50883
B#N

Specifications

Boron nitride
BN
boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r
PZNSFCLAULLKQX-UHFFFAOYSA-N
boranylidyneamine
66227
24.82
99.5%
5 pc.
10043-11-5
MFCD00011317
Insoluble in water.
B#N
24.82
CHEBI:50883
Hot pressed rod,6.35mm dia x 406mm long
Bag
Videos
SDS Documents
Certificates