missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Boron 10, 10B, plasma standard solution, Specpure™, 10B 100μg/mL

Brand:  Alfa Aesar™ 045530.AD

Code : NEW

Additional Details : Weight : 0.00050kg Transport : UN number : 3264 Chem class : 1a Pack group : III

Quantity 50mL

Product Code. 15426995

  • £128.90 / 50mL


Estimated Shipment date
from Supplier 26-03-2018
Add to basket

Specifications

Specifications

Boron 10, 10B, plasma standard solution
Specpure™
MFCD00143869
1.0g/mL
50mL
10B 100μg/mL
Liquid
UN3264
100°C
Matrix: 2% HNO3
Videos
SDS